Learn More
Med Vet International Bone Plate Holding Forceps, 6in, J0212P, Each
Catalog No. 5000585189
Encompass_Preferred
These forceps are designed to maintain plate position through a hole in the plate. The ball, just behind the point of the jaw, locates in bone plate screw hole causing minimal interference elsewhere in the operation site. Also, useful as a reduction forceps in soft young bone, as the balls prevent over-penetration of the bone. 6" (15 cm).
Catalog No. 50-005-85189 Supplier Med Vet International Supplier No. JOR212P
These forceps are designed to maintain plate position through a hole in the plate. The ball, just behind the point of the jaw, locates in bone plate screw hole causing minimal interference elsewhere in the operation site. Also, useful as a reduction forceps in soft young bone, as the balls prevent over-penetration of the bone. 6" (15 cm).