Learn More
Med Vet International Bone Plate, Round Holes, J0403B, 2.0mm, 4 holes, 20mm, Each
Catalog No. 5000582805
Encompass_Preferred
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.
Catalog No. 50-005-82805 Supplier Med Vet International Supplier No. JOR403B
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.