Learn More
Med Vet International Bone Plate, Round Holes, J0403D, 2.0mm, 5 holes, 25mm, Each
Catalog No. 5000582699
Encompass_Preferred
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.
Catalog No. 50-005-82699 Supplier Med Vet International Supplier No. JOR403D
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.