Learn More
Med Vet International Bone Plate, Round Holes, J0403E, 2.0mm, 5 holes, 28mm, Each
Supplier:  Med Vet International JOR403E
Encompass_Preferred
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.
Catalog No. 50-005-82625