Learn More
Med Vet International Bone Plate, Round Holes, J0403G, 2.0mm, 6 holes, 33mm, Each
Catalog No. 5000582746
Encompass_Preferred
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.
Catalog No. 50-005-82746 Supplier Med Vet International Supplier No. JOR403G
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.