Learn More
Med Vet International Bone Plate, Round Holes, J0403H, 2.0mm, 7 holes, 35mm, Each
Catalog No. 5000582050
Encompass_Preferred
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.
Catalog No. 50-005-82050 Supplier Med Vet International Supplier No. JOR403H
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.