Learn More
Med Vet International Bone Plate, Round Holes, J0403I, 2.0mm, 7 holes, 38mm, Each
Supplier: Med Vet International JOR403I
Available in a wide range of sizes, the 2mm plates used with either 2mm self-tapping screws or 2mm cortical screws provide for the secure repair of the small bones of a dog or cat.