Learn More
Cosmo Bio Usa Inc Bone Resorption Assay Kit - CSR-BRA-S96X2KIT, 2 x 96 strip-well rxns
SDP
Catalog No. NC3351462
Encompass
Bone Resorption Assay Kit is for measurement of cellular bone resorption activity.Bone resorption activity releases fluoresceinamine-labeled chondroitin sulfate from plate-bound calcium phosphate and is quantitated by spectrophotometric evaluation of culture media fluorescence intensity.
Catalog No. NC3351462 Supplier Cosmo Bio Usa Inc Supplier No. CSRBRAS96X2KIT
Bone Resorption Assay Kit is for measurement of cellular bone resorption activity.Bone resorption activity releases fluoresceinamine-labeled chondroitin sulfate from plate-bound calcium phosphate and is quantitated by spectrophotometric evaluation of culture media fluorescence intensity.