Learn More
Fine Science Tools BONE SCISSORS TUNGSTEN CARBIDE
SDP
Catalog No. NC0618469
Encompass
Tip Shape: Straight/ Cutting Edge: 15mm/ Alloy / Material: Stainless Steel/ Length: 16cm/ Joint Action: Single/ Feature: Tungsten Carbide
Catalog No. NC0618469 Supplier Fine Science Tools Supplier No. 1603016
Tip Shape: Straight/ Cutting Edge: 15mm/ Alloy / Material: Stainless Steel/ Length: 16cm/ Joint Action: Single/ Feature: Tungsten Carbide