Learn More
Electron Microscopy Sciences Bone Shears Stainless Steel, , 8" OAL
SDP
Catalog No. 5024776
Encompass_Preferred
Made from stainless steel with a removable bar spring; perfect for cutting cartilage and bone.  
OAL: 8"
Catalog No. 50-247-76 Supplier Electron Microscopy Sciences Supplier No. 7289408
Made from stainless steel with a removable bar spring; perfect for cutting cartilage and bone.  
OAL: 8"