Learn More
Enzo Life Sciences Bone sialoprotein polyclonal antibody (20µl)
SDP
Supplier:  Enzo Life Sciences ALX210312R020
Encompass_Preferred
Host: Rabbit. Immunogen: Purified human bone sialoprotein (bone extract). UniProt ID: P21815. Species reactivity: Human, Rat. Creossreactivity: Does not cross-react with human osteopontin or osteonectin. Applications: ELISA, ICC, IHC (PS), WB. Recommended Dilutions: Immunocytochemistry (1:100) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Lyophilized. Contains no preservatives. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8106