Learn More
BioVendor, LLC Bone Slices For 96W Plate 50Pk BioVendor LLC
SDP
Supplier:  BioVendor, LLC DT1BON100096
Encompass
TypeMediumDescriptionBone slices are for accurate in vitro assessment of osteoclastic bone resorption either by traditional staining or by use of the CrossLaps for Culture (CTX-I) ELISA assay.The bone slices are made from the cortical part of the femur of bovine bones, and are perfectly fitted for the 96 well format. Bone slices permit the in vitro quantification of bone resorption using the Crosslaps for Culture (CTX-I) ELISA as cultured osteoclasts, upon resorption, release collagen type-I fragment CTX to the osteoclast culture supernatant.Storage/ExpirationThe bone slices are stored in 70% ethanol at 4¿C.
Catalog No. 50-196-3846