Learn More
Med Vet International Bone Tunnel Borer, 2.7mm, J0236T
Catalog No. 5000584383
Encompass_Preferred
The borer is a small handle with an attached drill bit. This hand instrument is used to create a bone tunnel quickly and easily without having to set up a complete orthopedic drill (i.e., CCL repair through tibial tuberosity). Stainless steel. 18cm.
Catalog No. 50-005-84383 Supplier Med Vet International Supplier No. JOR236T
The borer is a small handle with an attached drill bit. This hand instrument is used to create a bone tunnel quickly and easily without having to set up a complete orthopedic drill (i.e., CCL repair through tibial tuberosity). Stainless steel. 18cm.