Learn More
Med Vet International Bone Twist Drill, J0372K, 2.3mm, Each
Catalog No. 5000581523
Encompass_Preferred
For pre-drilling positive profile pins. Pre-drill with a bit slightly smaller than the pin shank.
Catalog No. 50-005-81523 Supplier Med Vet International Supplier No. JOR372K
For pre-drilling positive profile pins. Pre-drill with a bit slightly smaller than the pin shank.