Learn More
Med Vet International Bone Twist Drill, J0372L, 3mm, Each
Catalog No. 5000582914
Encompass_Preferred
For pre-drilling positive profile pins. Pre-drill with a bit slightly smaller than the pin shank.
Catalog No. 50-005-82914 Supplier Med Vet International Supplier No. JOR372L
For pre-drilling positive profile pins. Pre-drill with a bit slightly smaller than the pin shank.