Learn More
World Precision Instrument Bone Wax
SDP
Catalog No. 50822813
Encompass
Achieve homeostasis of bone by applying a small amount of sterile bone wax. The wax is a combination of beeswax, isopropyl palmitate and mineral oil, which stops bleeding at the site.
Catalog No. 50-822-813 Supplier World Precision Instrument Supplier No. 501771
Achieve homeostasis of bone by applying a small amount of sterile bone wax. The wax is a combination of beeswax, isopropyl palmitate and mineral oil, which stops bleeding at the site.