Learn More
Braintree Scientific Bone Wax, Sterile Synthetic, 12/box
SDP
Catalog No. 501954681
Encompass
Bone Wax Bone wax is used to control bleeding in procedures that involve cutting through bones, such as thoracic, neurosurgery, orthopedic, oral and maxillofacial surgeries. Soft and easy to form and apply. Each easy-to-open foil package contains 2.5 grams of wax. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4681 Supplier Braintree Scientific Supplier No. DYNJBW26
Bone Wax Bone wax is used to control bleeding in procedures that involve cutting through bones, such as thoracic, neurosurgery, orthopedic, oral and maxillofacial surgeries. Soft and easy to form and apply. Each easy-to-open foil package contains 2.5 grams of wax. **Sold for research purposes only and not for human use**