Learn More
BioVendor, LLC BoneTRAP (Human TRAcP 5b) ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC SBTR201R
Encompass
BoneTRAP (Human TRAcP 5b) ELISA (96 wells). Immunoassay test kit for quantitative determination of active isoform 5b of the tartrate-resistant acid phosphatase (TRACP). TypeImmunocapture enzyme activity assayApplicationsSerum, Plasma-EDTASample Requirements100 µl/wellShippingShipped on blue ice packs. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box).Calibration Range0.5 - 10 U/LLimit of Detection< 0.5 U/L
Catalog No. NC1885860