Learn More
Roboz Surgical BONN CURVED SCISSORS
SDP
Catalog No. NC9503771
Encompass
Bonn Scissors; Curved; Sharp Points; 15mm Blade Length; 3-1/2 in. Overall Length
Catalog No. NC9503771 Supplier Roboz Surgical Supplier No. RS5841
Bonn Scissors; Curved; Sharp Points; 15mm Blade Length; 3-1/2 in. Overall Length