Learn More
Stoelting™ Bonn Scissors, Tungsten
SDP
Catalog No. 10001488 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Straight, S/S tip
Straight, Strong cut
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-488 Straight, S/S tip
10-000-508 Straight, Strong cut
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-488 Supplier Stoelting™ Supplier No. 5213207
Add to Cart
Add to Cart
Tungsten carbide scissors are especially good for cutting cartilage or for many repeated cuts such as in necropsy studies.
  • Tungsten carbide is 5 times harder than stainless steel.
  • Scissors with tungsten carbide inserts along their blades last much longer than stainless steel scissors.
Specifications
For Use With (Application) Neuroscience
Length (English) 3.5 in.
Material Tungsten
Width (Metric) 15 mm blades
Model Straight, S/S tip