Learn More
VAI Cleanroom Boot Covers
Catalog No. 19547222 Shop All VAI Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-547-222 Large
19-547-223 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-547-222 Supplier VAI Supplier No. 150SCNS1520
Add to Cart
Add to Cart
This item is not returnable. View return policy
Boot covers are latex-free and lint-free with a seamless bottom. Suitable for use in cleanroom environments.
  • Individually inspected for size, stitching, and workmanship
  • Neatly stacked and cleanroom packaged
  • Low linting fabric
Specifications
Size Large
Color Gray
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Non-skid
Packaging Quantity 100 Pairs/Cs.
Height (English) 7.75 in.
Height (Metric) 197 mm
Product Type Controlled Environments Boot Covers