Learn More
Protexer Corporation Shoe Cover Refill - CPE - Case - 525 pairs
Catalog No. NC0904462
Encompass_Preferred
Constructed of a durable spunbond polypropylene base-layer and coated with a heavy-duty layer of CPE. The blue CPE coating comes nearly all the way to the top of the bootie, making them even more durable and waterproof. The waterproof CPE layer, extra-deep design, and durable CPE coating make this bootie an ideal solution for users with large work boots. One size fits most (up to men’s size 15). Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.
Catalog No. NC0904462 Supplier Protexer Corporation Supplier No. KBCP525
Constructed of a durable spunbond polypropylene base-layer and coated with a heavy-duty layer of CPE. The blue CPE coating comes nearly all the way to the top of the bootie, making them even more durable and waterproof. The waterproof CPE layer, extra-deep design, and durable CPE coating make this bootie an ideal solution for users with large work boots. One size fits most (up to men’s size 15). Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.