Learn More
Apexbio Technology LLC BOP reagent 56602-33-6 100g
SDP
Catalog No. 5000093244
Encompass_Preferred
BOP reagent is a peptide-coupling reagent designed to facilitate amide bond formation between amino acids BOP reagent acts by activating carboxyl groups on amino acid derivatives or peptides promoting coupling reactions under mild conditions The mechanism involves the generation of benzotriazole group-bearing intermediates that enable efficient coupling with amino-functionalized reactants Based on these chemical properties BOP reagent is utilized in peptide synthesis and in the preparation of phenyl ester derivatives of amino acids for research in biochemical and pharmaceutical fields
Catalog No. 50-000-93244 Supplier Apexbio Technology LLC Supplier No. A7015100000
BOP reagent is a peptide-coupling reagent designed to facilitate amide bond formation between amino acids BOP reagent acts by activating carboxyl groups on amino acid derivatives or peptides promoting coupling reactions under mild conditions The mechanism involves the generation of benzotriazole group-bearing intermediates that enable efficient coupling with amino-functionalized reactants Based on these chemical properties BOP reagent is utilized in peptide synthesis and in the preparation of phenyl ester derivatives of amino acids for research in biochemical and pharmaceutical fields