Learn More
Electron Microscopy Sciences B.O.P. Biopsy Organizer Plate Table Top Light Box
SDP
Catalog No. 5033449
Encompass_Preferred
Provides bright, even light for illuminating specimen containers via the 8" x 10" illumination area. Runs on “D" cell batteries or an AC Adapter (sold separately).
Catalog No. 50-334-49 Supplier Electron Microscopy Sciences Supplier No. 6392050
Provides bright, even light for illuminating specimen containers via the 8" x 10" illumination area. Runs on “D" cell batteries or an AC Adapter (sold separately).