Learn More
Aviva Systems Biology Corp BOP1 Antibody - N-terminal region (ARP46225_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46225_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BOP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BOP1. Alternate names: KIAA0124ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59276