Learn More
AirSea USA Biopack 1 - 330ml UN Combination Packaging, 4G/Class 6.2
Catalog No. 501534818
Encompass_Preferred
Biopack 1 330ml Combination packaging complete with one 330ml biotube. The 330ml biotube is capable of withstanding 110kPa, is fully autoclavable and comes with appropriate hazard labels, absorbent cushioning and instructions. Biopak 1 is also approved with four 50ml Biotube50's. (Code 334)
Catalog No. 50-153-4818 Supplier AirSea USA Supplier No. 332
Biopack 1 330ml Combination packaging complete with one 330ml biotube. The 330ml biotube is capable of withstanding 110kPa, is fully autoclavable and comes with appropriate hazard labels, absorbent cushioning and instructions. Biopak 1 is also approved with four 50ml Biotube50's. (Code 334)