Learn More
Enzo Life Sciences Bopindolol malonate (100mg). CAS: 82857-38-3
SDP
Catalog No. 502010321
Encompass_Preferred
A slowly dissociating β1-adrenergic receptor antagonist. Antihypertensive. Antioxidant (IC50=1.8 µM) and free radical scavenging activity. Purity: ≥97% (HPLC). Solubility: Soluble in water at 3mg/ml. Long Term Storage: Ambient.
Catalog No. 50-201-0321 Supplier Enzo Life Sciences Supplier No. BMLAR1000100
A slowly dissociating β1-adrenergic receptor antagonist. Antihypertensive. Antioxidant (IC50=1.8 µM) and free radical scavenging activity. Purity: ≥97% (HPLC). Solubility: Soluble in water at 3mg/ml. Long Term Storage: Ambient.