Learn More
Cayman Chemical Bopindolol (malonate), Purity: ≥98%, CAS: 82857-38-3, 100 mg
SDP
Catalog No. 502993755
Encompass_Preferred
A β-adrenergic receptor antagonist; decreases heart rate and blood pressure and attenuates ischemia-induced myocardial acidosis in dogs
Catalog No. 50-299-3755 Supplier Cayman Chemical Supplier No. 23365100 MG
A β-adrenergic receptor antagonist; decreases heart rate and blood pressure and attenuates ischemia-induced myocardial acidosis in dogs