Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane - 5-Ethyl-2-methylpyridine Complex 90.0+%, TCI America™
SDP

Catalog No. B408425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B408425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B408425ML Supplier TCI America Supplier No. B408425ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1014979-56-6
Molecular Formula C8H11BN
Molecular Weight (g/mol) 131.99
MDL Number MFCD16621137
InChI Key NHLPUHQNHDWXBY-UHFFFAOYSA-N
Synonym Borane - 5-Ethyl-2-picoline Complex, 5-Ethyl-2-methylpyridine Borane, 5-Ethyl-2-picoline Borane
PubChem CID 86280291
IUPAC Name (5-ethyl-2-methylpyridin-1-ium-1-yl)-λ¹-boranuidyl
SMILES [B-][N+]1=C(C)C=CC(CC)=C1

Specifications

Color Yellow
Quantity 25 mL
UN Number 2810
Formula Weight 135.02
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Borane - 5-Ethyl-2-methylpyridine Complex
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.