Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Borane-Ammonia Complex

Catalog No. 2877171G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-771-71G 1 g
28-771-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-771-71G Supplier Sigma Aldrich Supplier No. 2877171G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Borazane acts as a mild, efficient, and stereoselective reducing agent for aldehydes and ketones in protic or nonprotic media.It can be used as a storage medium for release of hydrogenBorazane is popularly used as an ammonia borane (AB) source for the formation of hexagonal boron nitride monolayers which are used as a substrate for graphene growth.

Specifications

CAS 13774-81-7
Molecular Formula H6BN
Linear Formula H3N -+ BH3
MDL Number MFCD00075639
Quantity 1 g
Synonym Amminetrihydroboron; Borazane
Molecular Weight (g/mol) 30.87 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.