Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane - Ammonia Complex 90.0+%, TCI America™
SDP

Catalog No. B508225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B508225G 25 g
B50825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B508225G Supplier TCI America Supplier No. B508225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13774-81-7
Molecular Formula BH3N
Molecular Weight (g/mol) 27.841
MDL Number MFCD00075639
InChI Key JBANFLSTOJPTFW-UHFFFAOYSA-N
Synonym Ammonia Borane
PubChem CID 6332567
IUPAC Name azane;boron
SMILES [B].N

Specifications

Melting Point 98°C
Color White
Quantity 25 g
Formula Weight 30.87
Percent Purity ≥90.0% (V)
Physical Form Crystalline Powder
Chemical Name or Material Borane - Ammonia Complex
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.