Learn More
Cambridge Isotope Laboratories DEUTERIUM (D, 99.9%) (D2,99.8%+HD,0.2%), 25 L, 7782-39-0
Catalog No. 501630872
Encompass
DEUTERIUM (D, 99.9%) (D2,99.8%+HD,0.2%), 25 L, 7782-39-0 **This item is shipped in a Code C stainless steel lecture bottle. The fee for the bottle will need added to the order as a separate charge, per each. Please include part# NC9763154 to the order.
Catalog No. 50-163-0872 Supplier Cambridge Isotope Laboratories Supplier No. DLM132925
DEUTERIUM (D, 99.9%) (D2,99.8%+HD,0.2%), 25 L, 7782-39-0 **This item is shipped in a Code C stainless steel lecture bottle. The fee for the bottle will need added to the order as a separate charge, per each. Please include part# NC9763154 to the order.