Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-di-tert-butylphosphine complex, 98%, Thermo Scientific™

Catalog No. AAH2689903
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2689903 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2689903 Supplier Thermo Scientific Chemicals Supplier No. H2689903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 128363-76-8
Molecular Formula C8H19BP
Linear Formula [(CH3)3C]2PH·BH3
Quantity 1g
MDL Number MFCD03412071
UN Number UN2813
Sensitivity Moisture sensitive
Synonym di-tert-butylphosphane boranylidyne, borane di tert-butyl phosphine complex, borane-di tert-butyl phosphine complex, di-t-butylphosphine-borane, di-tert-butylphosphine borane, di-tert-butylphosphane borane, di-tert-butylphosphine trihydroboron, borane di-tert-butylphosphine complex, boron-di-tert-butylphosphane 1/1
InChI Key NFTWUNMDOWXENK-UHFFFAOYSA-N
SMILES [B].CC(C)(C)PC(C)(C)C
IUPAC Name boron;ditert-butylphosphane
Molecular Weight (g/mol) 157.024
PubChem CID 22572032
Formula Weight 160.05
Percent Purity 98%
Chemical Name or Material Borane-di-tert-butylphosphine complex
Show More Show Less
Hazard Category H261
Hazard Statement GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.