Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Borane Dimethyl Sulfide Complex

Catalog No. 503081437 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
4 x 25 mL
800 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1437 25 mL
17-982-5100ML 100 mL
17-982-525ML 4 x 25 mL
17-982-5800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 50-308-1437 Supplier Sigma Aldrich Supplier No. 17982525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Borane dimethyl sulfide complex (BMS) is a commonly used reagent and a mediator for hydroboration reaction for the preparation of organoborane compounds, which are utilized as key intermediates in organic synthesis. BMS is also employed as a reducing agent for the reduction of various functional groups such as aldehydes, ketones, epoxides, esters, and carboxylic acids to corresponding alcohols.Borane-dimethyl sulfide (BH3 Me2S) can be used as a reagent: For the selective synthesis of 1,3,5-oxygenated compounds from dimethyl 3-oxoglutarate. For the conversion of ozonides to alcohols. In the CBS-catalyzed asymmetric reduction of ferrocenyl-1,3-diketones to 1,3-diols. For enantioselective reduction of ketones to chiral secondary alcohols in the presence of C3-symmetric tripodal hydroxyamide as a ligand. For the hydroboration reduction and other applications. With a dendrimeric supported L-pyrrolidinol in the asymmetric reduction of indanones and tetralones.10.0-10.2 M as BH3. May contain excess methyl sulfide.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 13292-87-0
Density 0.801 g/mL at 25°C
Molecular Formula C2H9BS
Linear Formula (CH3)2S -+ BH3
Quantity 25 mL
MDL Number MFCD00013189
Synonym (Dimethyl sulfide)trihydroboron; BMS; Borane-dimethyl sulfide
Molecular Weight (g/mol) 75.97 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.