Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Borane Dimethyl Sulfide Complex Solution

Catalog No. 775118100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-511-8100ML 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 77-511-8100ML Supplier Sigma Aldrich Supplier No. 775118100ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 1.0 M in 2-methyltetrahydrofuran aligns with Safer Solvents and Auxiliaries, Use of Renewable Feedstocks and Inherently Safer Chemistry for Accident Prevention. Find details here.Reactant used as a regioselective reducing agentReactant involved in: Hydroboration / oxidationSynthesis of cage compoundsSure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 105299-45-4
Melting Point 270°C to 280°C
Molecular Formula C13H10N2O3
Linear Formula C13H10N2O3
MDL Number MFCD00225268
Quantity 250 mg
Synonym BMS; Trihydro[thiobis[methane]]boron
Molecular Weight (g/mol) 242.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.