Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Borane Dimethyl Sulfide Complex Solution

Catalog No. 192120800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-212-0800ML 800 mL
19-212-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 19-212-0800ML Supplier Sigma Aldrich Supplier No. 192120800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Borane dimethyl sulfide complex (BMS) acts as a highly efficient and selective reducing agent in the presence of catalytic sodium tetrahydroborate for +a-hydroxy esters. Asymmetric borane reduction of a variety of prochiral ketones with BMS using spiroborate esters as catalyst has been reported.Reactant used as a regioselective reducing agentReactant involved in: Hydroboration / oxidationSynthesis of cage compounds

Specifications

CAS 13292-87-0
Density 0.855 g/mL at 25°C
Molecular Formula C2H9BS
Linear Formula (CH3)2S -+ BH3
MDL Number MFCD00013189
Quantity 800 mL
Synonym BMS; Trihydro[thiobis[methane]]boron
Molecular Weight (g/mol) 75.97 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.