Learn More
Strem, An Ascensus Company CAS# 13292-87-0. 25g. Borane, dimethylsulfide complex (contains 5-6% excess dimethylsulfide). MFCD00013189
SDP
Catalog No. 5090108675
Encompass
CAS# 13292-87-0. 25g. Borane, dimethylsulfide complex (contains 5-6% excess dimethylsulfide). MFCD00013189. Molecular Weight: 75.97. Molecular Formula: (CH3)2S:BH3. Color/form: colorless liq. Strem# 05-0110. http://www.strem.com/catalog/v/05-0110/
Catalog No. 50-901-08675 Supplier Strem, An Ascensus Company Supplier No. 05011025G
CAS# 13292-87-0. 25g. Borane, dimethylsulfide complex (contains 5-6% excess dimethylsulfide). MFCD00013189. Molecular Weight: 75.97. Molecular Formula: (CH3)2S:BH3. Color/form: colorless liq. Strem# 05-0110. http://www.strem.com/catalog/v/05-0110/