Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Borane-protected tris(2-carboxyethyl)phosphine | 188714-28-5 | 99.5% | 296.11 g/mol | C14H22BO4P | 5 MG
SDP

Catalog No. 5000221097
Encompass_Preferred

PB1 is a borane-protected analogue of tris(2-carboxyethyl)phosphine (TCEP) that functions as a cell-permeable disulfide reducing reagent for biochemical and cellular research. The compound is characterized by high purity, DMSO solubility, and defined storage and stability recommendations for reliable experimental use.

  • Cell-permeable disulfide reducing agent.
  • Borane-protected analogue offering improved stability.
  • High purity (≈99.5%) for reproducible results.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation may be needed.
  • Recommended storage at 4°C under inert gas; solution stability at -80°C (6 months) or -20°C (1 month).
  • Suitable for biochemical and cell-based assays.

Catalog No. 50-002-21097 Supplier Medchemexpress LLC Supplier No. HY1386485MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PB1 is a borane-protected analogue of tris(2-carboxyethyl)phosphine (TCEP) that functions as a cell-permeable disulfide reducing reagent for biochemical and cellular research. The compound is characterized by high purity, DMSO solubility, and defined storage and stability recommendations for reliable experimental use.

  • Cell-permeable disulfide reducing agent.
  • Borane-protected analogue offering improved stability.
  • High purity (≈99.5%) for reproducible results.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation may be needed.
  • Recommended storage at 4°C under inert gas; solution stability at -80°C (6 months) or -20°C (1 month).
  • Suitable for biochemical and cell-based assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.