Learn More
Medchemexpress LLC Borane-protected tris(2-carboxyethyl)phosphine analogue | 188714-28-5 | 99.5% | C14H22BO4P | 10MG
SDP
Catalog No. 5000261221
Encompass_Preferred
PB1 is a borane-protected tris(2-carboxyethyl)phosphine analogue that serves as a potent intracellular disulfide reducing agent. It exhibits good cell permeability and intracellular stability, and has been reported to improve retinal ganglion cell survival after axotomy in vitro at nanomolar and picomolar concentrations, making it suitable for neuroprotection and intracellular redox studies.
  • Potent intracellular disulfide reducing agent.
  • Borane-protected tris(2-carboxyethyl)phosphine analogue.
  • Good cell permeability and intracellular stability.
  • Capable of forming a high intracellular concentration gradient.
  • Demonstrated neuroprotective effects in retinal ganglion cell assays at nM and pM ranges.
  • Provided as a research-grade powder for biochemical applications.
Catalog No. 50-002-61221 Supplier Medchemexpress LLC Supplier No. HY13864810MG
PB1 is a borane-protected tris(2-carboxyethyl)phosphine analogue that serves as a potent intracellular disulfide reducing agent. It exhibits good cell permeability and intracellular stability, and has been reported to improve retinal ganglion cell survival after axotomy in vitro at nanomolar and picomolar concentrations, making it suitable for neuroprotection and intracellular redox studies.
  • Potent intracellular disulfide reducing agent.
  • Borane-protected tris(2-carboxyethyl)phosphine analogue.
  • Good cell permeability and intracellular stability.
  • Capable of forming a high intracellular concentration gradient.
  • Demonstrated neuroprotective effects in retinal ganglion cell assays at nM and pM ranges.
  • Provided as a research-grade powder for biochemical applications.