Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-pyridine complex, 95%, (ca. 9.4M), AcroSeal™

Catalog No. AC427131000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427131000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427131000 Supplier Thermo Scientific Chemicals Supplier No. 427131000
  • $239.00
    $203.65 / Each of 1

    Save $35.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 110-51-0 | C5H8BN | 92.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-51-0
Molecular Formula C5H8BN
Molecular Weight (g/mol) 92.94
MDL Number MFCD00012435
InChI Key LPGWNCNRGQANGC-UHFFFAOYSA-N
Synonym Pyridineborane
IUPAC Name (pyridin-1-ium-1-yl)boranuide
SMILES [BH3-][N+]1=CC=CC=C1

Specifications

Melting Point 10.0°C to 11.0°C
Color Amber to Colorless
Density 0.9200g/mL
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.5300 to 1.5320
Quantity 100 mL
Specific Gravity 0.92
Solubility Information Solubility in water: reacts slowly with water. Other solubilities: soluble in methanol,ethanol
Viscosity 2.5 mm2/s (22°C)
Formula Weight 92.94
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Borane-pyridine complex, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
EINECSNumber 203-773-4
Recommended Storage Product may darken during storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.