Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-pyridine complex, 95%

Catalog No. AAL1317809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1317809 10 g
AAL1317818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1317809 Supplier Thermo Scientific Chemicals Supplier No. L1317809
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 110-51-0 | C5H8BN | 92.94 g/mol

Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

Solubility
It hydrolyzes slowly in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-51-0
Molecular Formula C5H8BN
Molecular Weight (g/mol) 92.94
MDL Number MFCD00012435
InChI Key LPGWNCNRGQANGC-UHFFFAOYSA-N
Synonym Pyridine-borane
IUPAC Name (pyridin-1-ium-1-yl)boranuide
SMILES [BH3-][N+]1=CC=CC=C1

Specifications

Melting Point 9°C to 11°C
Density 0.92
Boiling Point 65°C (1 mmHg)
Flash Point 73°C (163°F)
Odor Unpleasant
Refractive Index 1.532
Quantity 10 g
UN Number UN2810
Beilstein 3674913
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes slowly in water.
Formula Weight 92.93
Percent Purity 95%
Chemical Name or Material Borane-pyridine complex
Show More Show Less
Hazard Category H227-H301-H310+H330-H351
Hazard Statement GHS H Statement
H301-H310-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P235-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P308+P313-P310-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-773-4
RTECSNumber US3675000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.