Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Borane Tert-Butylamine Complex

Catalog No. 18021125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-021-125G 25 g
18-021-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-021-125G Supplier Sigma Aldrich Supplier No. 18021125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Borane Tert-Butylamine Complex

Specifications

CAS 7337-45-3
Melting Point 98°C to 100°C
Molecular Formula C4H14BN
Linear Formula (CH3)3CNH2 -+ BH3
MDL Number MFCD00075635
Quantity 25 g
Synonym tert-Butylamine borane; NSC 114045; Trihydro(2-methyl-2-propanamine) boron
Molecular Weight (g/mol) 86.97 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.