Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-triphenylphosphine complex, 97%, Thermo Scientific™

Catalog No. AA4296906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4296906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4296906 Supplier Thermo Scientific Chemicals Supplier No. 4296906

missing translation for 'validMainframeMsg'

In hydroboration reactions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2049-55-0
Molecular Formula C18H15BP
Molecular Weight (g/mol) 273.102
MDL Number MFCD00012427
InChI Key QFLQJPFCNMSTJZ-UHFFFAOYSA-N
Synonym triphenylphosphine borane, borane triphenylphosphine complex, triphenylphosphineborane, boron; triphenylphosphane, triphenylphosphinborane, pubchem18246, acmc-209fav, borane-triphenylphosphine complex, triphenylphosphine boranylidyne
PubChem CID 6364612
IUPAC Name boron;triphenylphosphane
SMILES [B].C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 189°C to 191°C
Color White
Odor Unpleasant
Linear Formula (C6H5)3P·BH3
Quantity 5 g
UN Number UN3396
Sensitivity Air & Moisture Sensitive
Formula Weight 276.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Borane-triphenylphosphine complex
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H261-H228-H315-H319-H335
In contact with water releases flammable gas.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P231+P232-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.