Learn More
Medchemexpress LLC Borate, 5-[(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-1H-pyrrole-2-pentanoato difluoro | 217075-24-6 | 99.7% | 320.14 g/mol | C16H19BF2N2O2 | 10 MG
SDP
Catalog No. 5000298852
Encompass_Preferred
BODIPY FL C5 is a green fluorescent fatty acid probe used for labeling lipids and membranes and as a precursor for synthesizing fluorescent phospholipids. It fluoresces in both aqueous and lipid environments and is supplied as an orange to red solid at high purity for research applications. Not for drug or household use.
  • High purity (99.7%).
  • Fluoresces in aqueous and lipid environments.
  • Suitable for membrane labeling and lipid trafficking assays.
  • Orange to red solid, easy to handle.
  • Supplied in 10 mg quantities for laboratory workflows.
  • Includes COA and SDS for quality and safety information.
Catalog No. 50-002-98852 Supplier Medchemexpress LLC Supplier No. HYD161010MG
BODIPY FL C5 is a green fluorescent fatty acid probe used for labeling lipids and membranes and as a precursor for synthesizing fluorescent phospholipids. It fluoresces in both aqueous and lipid environments and is supplied as an orange to red solid at high purity for research applications. Not for drug or household use.
  • High purity (99.7%).
  • Fluoresces in aqueous and lipid environments.
  • Suitable for membrane labeling and lipid trafficking assays.
  • Orange to red solid, easy to handle.
  • Supplied in 10 mg quantities for laboratory workflows.
  • Includes COA and SDS for quality and safety information.