Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Ricca Chemical
SDP

Catalog No. R1040000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1040000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1040000500 Supplier Ricca Chemical Company Supplier No. R1040000500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Analysis of Ammonia and organic Nitrogen

Specifications

Chemical Name or Material Borate Buffer
CAS 7732-18-5, 1303-96-4, 1310-73-2
CAS Min % 97, 0.46, 0.07
CAS Max % 100, 0.48, 0.07
Color Colorless
For Use With (Application) Nitrogen Analysis
pH 9.5
Physical Form Liquid
Quantity 500 mL
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.