Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Borate Buffer pH 9.5 APHA for Ammonia Nitrogen (1L)
SDP

Catalog No. NC9583769
Encompass_Preferred

Borate Buffer pH 9.5 APHA for Ammonia Nitrogen (1L)

Catalog No. NC9583769 Supplier Aqua Solutions Supplier No. 12801L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Borate Buffer pH 9.5 APHA for Ammonia Nitrogen (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.