Learn More
Boston Bioproducts Inc Borate Buffer (0.5 M, pH 8.5) - 500ml
Catalog No. NC9851660
Encompass
The Borate Buffer is an isotonic buffer essential for maintaining stable pH levels in biochemical processes. Typically operating within a pH range of 8.0 to 9.5, this buffer supports several applications including enzymatic assays, protein purification, as a DNA electrophoresis buffer, in coating procedures, and for coupling proteins. The Borate Buffer is also used as plant tissue culture media, and has a strong bactericidal effect.
Catalog No. NC9851660 Supplier Boston Bioproducts Inc Supplier No. BB66500ML
The Borate Buffer is an isotonic buffer essential for maintaining stable pH levels in biochemical processes. Typically operating within a pH range of 8.0 to 9.5, this buffer supports several applications including enzymatic assays, protein purification, as a DNA electrophoresis buffer, in coating procedures, and for coupling proteins. The Borate Buffer is also used as plant tissue culture media, and has a strong bactericidal effect.