Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borax 99% MP Biomedicals

Catalog No. ICN19030801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19030801 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19030801 Supplier MP Biomedicals, Inc Supplier No. 0219030801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
Synonym Sodium tetraborate

Specifications

Physical Form Powder
Quantity 1 kg
Assay Percent Range 99%
Formula Weight 381.4
Density 1.730 g/mL
Chemical Name or Material Borax
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.