Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borax Anhydrous MP Biomedicals

Catalog No. ICN19030980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19030980 100 g
ICN19030990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19030980 Supplier MP Biomedicals, Inc Supplier No. 0219030980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1330-43-4 | Formula: B4Na2O7 | Weight: 201.21 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
Synonym Sodium Tetraborate, Sodium Borate Anhydrous, Sodium Borate
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Density 2.367 g/mL
Formula Weight 201.2
Percent Purity 99.77%
Chemical Name or Material Borax, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.