Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Borax Tetra, Sodium Borate, Reagent 500 GR
SDP

Supplier:  Electron Microscopy Sciences 21130

Encompass_Preferred

(Sodium Tetraborate, Decahydrate)
Na₂B₄O₇ .₁₀H₂O F.W. 381.37
Assay: 99.5-105%
pH: 9.15-9.20 0.01M @ 25°C

Catalog No. 50-247-83


May include imposed supplier surcharges.
Edge
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.